Learn More
Waters Corp Method Validation Kit, PFP/Fluoro-Phenyl, 100 Å, 5 μm, 4.6 mm X 150 mm
Catalog No. 502392819 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a Method Validation Kit designed for chromatography applications. It features HSS particle technology and a PFP/Fluoro-Phenyl stationary phase, ensuring reliable performance in analytical procedures.
  • PFP/Fluoro-Phenyl stationary phase
  • 5 μm particle size
  • 100 Å pore size
  • 4.6 mm inner diameter
  • 150 mm length
  • Units per package: 3 pk
  • HSS particle technology
Catalog No. 50-239-2819 Supplier Waters Corp Supplier No. 186005899
This product is a Method Validation Kit designed for chromatography applications. It features HSS particle technology and a PFP/Fluoro-Phenyl stationary phase, ensuring reliable performance in analytical procedures.
  • PFP/Fluoro-Phenyl stationary phase
  • 5 μm particle size
  • 100 Å pore size
  • 4.6 mm inner diameter
  • 150 mm length
  • Units per package: 3 pk
  • HSS particle technology