Learn More
Waters Corp Waters XSelect HSS PFP XP Column, HSS PFP, 2.5 μm, 4.6 x 75 mm
Catalog No. 50201571 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These fluorinated phases offer beneficial alternatives to standard C18 phases, particularly when selectivity for bases or planar aromatic compounds is needed. They provide exceptional separation performance, robustness, and throughput for HPLC assays, and are fully compatible with all HPLC, UHPLC, and UPLC Technology platforms.
  • Offers beneficial alternatives to standard C18 phases, particularly for selectivity for bases or planar aromatic compounds.
  • Provides exceptional separation performance, robustness, and throughput.
  • Fully compatible with all HPLC, UHPLC, and UPLC technology platforms.
  • Designed to withstand higher pressures (up to 9000 psi for 4.6 mm ID and 18,000 psi for 2.1 mm and 3.0 mm ID).
  • Offers flexibility in choosing column length (30 mm, 50 mm, 75 mm, 100 mm, and 150 mm) to balance resolution and throughput.
Catalog No. 50-201-571 Supplier Waters Corp Supplier No. 186006182
Add to Cart
Add to Cart
These fluorinated phases offer beneficial alternatives to standard C18 phases, particularly when selectivity for bases or planar aromatic compounds is needed. They provide exceptional separation performance, robustness, and throughput for HPLC assays, and are fully compatible with all HPLC, UHPLC, and UPLC Technology platforms.
  • Offers beneficial alternatives to standard C18 phases, particularly for selectivity for bases or planar aromatic compounds.
  • Provides exceptional separation performance, robustness, and throughput.
  • Fully compatible with all HPLC, UHPLC, and UPLC technology platforms.
  • Designed to withstand higher pressures (up to 9000 psi for 4.6 mm ID and 18,000 psi for 2.1 mm and 3.0 mm ID).
  • Offers flexibility in choosing column length (30 mm, 50 mm, 75 mm, 100 mm, and 150 mm) to balance resolution and throughput.