Learn More
Waters Corp Waters Column, PFP, 2.5 μm, 2.1 x 75 mm
Supplier: Waters Corp 176002644
This HSS PFP column is a low coverage, unendcapped pentafluorophenyl phase. It offers exceptional separation of closely related compounds and provides alternate selectivity compared to traditional alkyl chain columns through an intricate combination of retention mechanisms. Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance (XP) columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, while also enabling a seamless transition path toward UPLC adoption.
- Chemistry: PFP/Fluoro-Phenyl
- Separation mode: Reversed phase
- Particle substrate: Silica
- Particle size: 2.5 μm
- Pore size: 100Å
- Inner diameter: 2.1 mm
- Length: 75 mm
- pH range: 2 to 8 pH
- Maximum pressure: 18000 psi (1240 Bar)
- Units per package: 3 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.