Learn More
Waters Corp Waters Column, PFP/Fluoro-Phenyl, 2.5 μm, 2.1 mm X 100 mm
Catalog No. 50179304 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS PFP column is a low coverage, unendcapped pentafluorophenyl phase designed to provide exceptional separation of closely related compounds. It offers alternate selectivity compared to traditional alkyl chain columns through a complex combination of retention mechanisms. The column is packed in ultra-low dispersion hardware, and its 2.5 μm eXtended Performance [XP] columns deliver outstanding separation performance, robustness, and throughput for HPLC assays, while also enabling a smooth transition to UPLC adoption.
  • PFP/Fluoro-Phenyl chemistry
  • Reversed phase separation
  • Silica particle substrate
  • pH range 2 to 8 pH
  • Maximum pressure 18000 psi (1240 Bar)
  • 2.5 μm particle size
  • 100 Å pore size
  • 2.1 mm inner diameter
  • 100 mm length
  • UPLC, UHPLC system compatible
  • eCord enabled
  • 3 pk per package
Catalog No. 50-179-304 Supplier Waters Corp Supplier No. 176002645
The XSelect HSS PFP column is a low coverage, unendcapped pentafluorophenyl phase designed to provide exceptional separation of closely related compounds. It offers alternate selectivity compared to traditional alkyl chain columns through a complex combination of retention mechanisms. The column is packed in ultra-low dispersion hardware, and its 2.5 μm eXtended Performance [XP] columns deliver outstanding separation performance, robustness, and throughput for HPLC assays, while also enabling a smooth transition to UPLC adoption.
  • PFP/Fluoro-Phenyl chemistry
  • Reversed phase separation
  • Silica particle substrate
  • pH range 2 to 8 pH
  • Maximum pressure 18000 psi (1240 Bar)
  • 2.5 μm particle size
  • 100 Å pore size
  • 2.1 mm inner diameter
  • 100 mm length
  • UPLC, UHPLC system compatible
  • eCord enabled
  • 3 pk per package