Learn More
Waters Corp Waters XSelect HSS PFP Column, 100Å, 2.5 μm, 2.1 mm x 100 mm
Catalog No. 50179563 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS PFP column is a low coverage, unendcapped pentafluorophenyl phase, providing exceptional separation and alternate selectivity for HPLC assays and UPLC adoption.
  • Exceptional separation of closely related compounds.
  • 2.5 μm eXtended Performance (XP) columns in ultra-low dispersion hardware.
  • Delivers robust performance and high throughput.
  • Seamless transition to UPLC adoption.
  • Dependable, reproducible, and scalable silica-based PFP LC columns.
  • HSS technology improves polar molecule retention and varied selectivity.
  • Unique phase for base and planar aromatic compound selectivity, increasing retention.
Catalog No. 50-179-563 Supplier Waters Corp Supplier No. 186006175
The XSelect HSS PFP column is a low coverage, unendcapped pentafluorophenyl phase, providing exceptional separation and alternate selectivity for HPLC assays and UPLC adoption.
  • Exceptional separation of closely related compounds.
  • 2.5 μm eXtended Performance (XP) columns in ultra-low dispersion hardware.
  • Delivers robust performance and high throughput.
  • Seamless transition to UPLC adoption.
  • Dependable, reproducible, and scalable silica-based PFP LC columns.
  • HSS technology improves polar molecule retention and varied selectivity.
  • Unique phase for base and planar aromatic compound selectivity, increasing retention.