Learn More
Waters Corp Waters Corporation Column, PFP, 100Å, 2.5 μm, 2.1 mm x 30 mm
Supplier: Waters Corp 186006172
The XSelect HSS PFP column is a low coverage, unendcapped pentafluorophenyl phase offering exceptional separation of closely related compounds and alternative selectivity to traditional alkyl chain columns. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, delivering excellent separation performance, robustness, and throughput for HPLC assays while facilitating a smooth transition to UPLC adoption.
- Chemistry: PFP/Fluoro-Phenyl
- Separation mode: Reversed Phase
- Particle substrate: Silica
- pH range: 2 to 8
- Maximum pressure: 18000 psi (1240 Bar)
- Endcapped: No
- Silanol activity: High
- Particle shape: Spherical
- Endfitting type: Parker-style
- System compatibility: UPLC, UHPLC
- USP classification: L43
- Carbon load: 7 %
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.