Learn More
Waters Corp XSelect HSS PFP Column, 100Å, 2.5 μm, 2.1 mm X 75 mm
Catalog No. 50179562 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS PFP column, a low coverage, unendcapped pentafluorophenyl phase, provides exceptional separation of closely related compounds. It offers alternate selectivity compared to traditional alkyl chain columns through intricate retention mechanisms. Packed in ultra-low dispersion hardware, its 2.5 μm eXtended Performance [XP] columns deliver robust performance and throughput for HPLC assays, facilitating UPLC adoption.
  • Low coverage, unendcapped pentafluorophenyl phase.
  • Provides exceptional separation of closely related compounds.
  • Offers alternate selectivity to traditional alkyl chain columns.
  • Utilizes an intricate combination of retention mechanisms.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance [XP] columns for robust performance.
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.
Catalog No. 50-179-562 Supplier Waters Corp Supplier No. 186006174
Add to Cart
Add to Cart
The XSelect HSS PFP column, a low coverage, unendcapped pentafluorophenyl phase, provides exceptional separation of closely related compounds. It offers alternate selectivity compared to traditional alkyl chain columns through intricate retention mechanisms. Packed in ultra-low dispersion hardware, its 2.5 μm eXtended Performance [XP] columns deliver robust performance and throughput for HPLC assays, facilitating UPLC adoption.
  • Low coverage, unendcapped pentafluorophenyl phase.
  • Provides exceptional separation of closely related compounds.
  • Offers alternate selectivity to traditional alkyl chain columns.
  • Utilizes an intricate combination of retention mechanisms.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance [XP] columns for robust performance.
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.