Learn More
Waters Corp Waters Corporation XSelect HSS PFP Fluoro-Phenyl XP Column, 2.5 μm, 3 x 100 mm
Catalog No. 50179567 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is a low coverage, unendcapped pentafluorophenyl phase designed for exceptional separation of closely related compounds. It offers alternate selectivity compared to traditional alkyl chain columns through an intricate combination of retention mechanisms, making it suitable for HPLC assays and providing a seamless transition path toward UPLC adoption.
  • Exceptional separation of closely related compounds.
  • Offers alternate selectivity for HPLC and UPLC assays.
  • Reliable, repeatable, and scalable silica-based PFP LC column.
  • High strength silica (HSS) technology enhances polar molecule retention and selectivity.
  • Mechanically strong particles sustain UPLC pressures.
  • Provides special phase selectivity for bases and planar aromatic compounds.
  • Recommended for higher retention of basic polar analytes.
Catalog No. 50-179-567 Supplier Waters Corp Supplier No. 186006179
This column is a low coverage, unendcapped pentafluorophenyl phase designed for exceptional separation of closely related compounds. It offers alternate selectivity compared to traditional alkyl chain columns through an intricate combination of retention mechanisms, making it suitable for HPLC assays and providing a seamless transition path toward UPLC adoption.
  • Exceptional separation of closely related compounds.
  • Offers alternate selectivity for HPLC and UPLC assays.
  • Reliable, repeatable, and scalable silica-based PFP LC column.
  • High strength silica (HSS) technology enhances polar molecule retention and selectivity.
  • Mechanically strong particles sustain UPLC pressures.
  • Provides special phase selectivity for bases and planar aromatic compounds.
  • Recommended for higher retention of basic polar analytes.