Learn More
Waters Corp XSelect Column, 100Å, 2.5 μm, 2.1 mm X 150 mm, 3/pk
Supplier:  Waters Corp 176002899
Encompass_Preferred
The XSelect HSS T3 is a low-coverage C18 phase compatible with 100% aqueous mobile phase, designed for developing separations for polar and non-polar compounds. These columns exhibit ultra-low MS bleed and are fully scalable and transferable to UPLC separations. Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance [XP] columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, providing a seamless transition path toward UPLC adoption.
  • Compatible with 100% aqueous mobile phase.
  • Exhibits ultra-low MS bleed.
  • Fully scalable and transferable to UPLC separations.
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.
  • Higher silanophilicity, aiding in retention of more polar compounds.
  • High transferability and outstanding reproducibility.
Catalog No. 50-465-939