Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XSelect Column, 100Å, 2.5 μm, 2.1 mm x 150 mm

Catalog No. 50466447 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XSelect HSS T3 Column features a low-coverage C18 phase, compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. These 2.5 μm eXtended Performance (XP) columns ensure exceptional separation performance and robustness for HPLC assays, offering ultra-low MS bleed and UPLC scalability.

  • Low-coverage C18 phase for polar and non-polar compound separation.
  • Ultra-low MS bleed.
  • UPLC scalable.
  • Manufactured in a cGMP ISO 9001:2000 certified factory.
  • High Strength Silica technology for unique selectivity, high analyte loading.
  • High silanophilicity ensures polar compound retention.
  • Handles pressures up to 18000 PSI (1240 Bar).
  • pH range of 2 to 8.
  • Usable up to 45°C.

Catalog No. 50-466-447 Supplier Waters Corp Supplier No. 186006739
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XSelect HSS T3 Column features a low-coverage C18 phase, compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. These 2.5 μm eXtended Performance (XP) columns ensure exceptional separation performance and robustness for HPLC assays, offering ultra-low MS bleed and UPLC scalability.

  • Low-coverage C18 phase for polar and non-polar compound separation.
  • Ultra-low MS bleed.
  • UPLC scalable.
  • Manufactured in a cGMP ISO 9001:2000 certified factory.
  • High Strength Silica technology for unique selectivity, high analyte loading.
  • High silanophilicity ensures polar compound retention.
  • Handles pressures up to 18000 PSI (1240 Bar).
  • pH range of 2 to 8.
  • Usable up to 45°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.