Learn More
Waters Corp Waters XSelect HSS T3 Method Validation Kit, 100Å, 2.5 μm, 2.1 mm X 150 mm, 3/pk
Catalog No. 50466489 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three columns packed with different sorbent batches. It features a low-coverage C18 phase, making it suitable for developing separations of both polar and non-polar compounds, and is compatible with 100% aqueous mobile phases. The columns offer ultra-low MS bleed and are fully scalable to UPLC separations, providing exceptional performance and robustness for HPLC assays with a clear path to UPLC adoption.
Features and Benefits:
  • Compatible with 100% aqueous mobile phase.
  • Exhibits ultra-low MS bleed.
  • Fully scalable and transferable to UPLC separations.
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.
  • Provides a seamless transition path toward UPLC adoption.
Catalog No. 50-466-489 Supplier Waters Corp Supplier No. 186006797
This method validation kit includes three columns packed with different sorbent batches. It features a low-coverage C18 phase, making it suitable for developing separations of both polar and non-polar compounds, and is compatible with 100% aqueous mobile phases. The columns offer ultra-low MS bleed and are fully scalable to UPLC separations, providing exceptional performance and robustness for HPLC assays with a clear path to UPLC adoption.
Features and Benefits:
  • Compatible with 100% aqueous mobile phase.
  • Exhibits ultra-low MS bleed.
  • Fully scalable and transferable to UPLC separations.
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.
  • Provides a seamless transition path toward UPLC adoption.