Learn More
Waters Corp WATERS CORPORATION XSelect HSS T3 Method Validation Kit, 2.5 μm, 100 Å, 4.6 mm, 150 mm
Catalog No. 50466491 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three columns packed with different sorbent batches. They feature a low-coverage C18 phase compatible with 100% aqueous mobile phases, ideal for separating polar and non-polar compounds. These columns offer ultra-low MS bleed and are fully scalable and transferable to UPLC separations. Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance [XP] columns ensure exceptional HPLC performance, robustness, and throughput, facilitating a smooth transition to UPLC.
  • Low-coverage C18 phase
  • Compatible with 100% aqueous mobile phase
  • Suitable for polar and non-polar compound separations
  • Ultra-low MS bleed
  • Fully scalable and transferable to UPLC
  • Ultra-low dispersion hardware
  • 2.5 μm XP columns
  • Exceptional HPLC performance, robustness, and throughput
  • Seamless transition to UPLC adoption
Catalog No. 50-466-491 Supplier Waters Corp Supplier No. 186006799
This method validation kit includes three columns packed with different sorbent batches. They feature a low-coverage C18 phase compatible with 100% aqueous mobile phases, ideal for separating polar and non-polar compounds. These columns offer ultra-low MS bleed and are fully scalable and transferable to UPLC separations. Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance [XP] columns ensure exceptional HPLC performance, robustness, and throughput, facilitating a smooth transition to UPLC.
  • Low-coverage C18 phase
  • Compatible with 100% aqueous mobile phase
  • Suitable for polar and non-polar compound separations
  • Ultra-low MS bleed
  • Fully scalable and transferable to UPLC
  • Ultra-low dispersion hardware
  • 2.5 μm XP columns
  • Exceptional HPLC performance, robustness, and throughput
  • Seamless transition to UPLC adoption