Learn More
Waters Corp Waters Corporation Column, C18, 3.5 um, 100 A, 1 mm x 50 mm
Catalog No. 50179712 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The trifunctional alkyl C18 bonded phase used for the HSS T3 sorbents is compatible with 100% aqueous mobile phase and provides ultra-low MS bleed, while promoting superior polar compound retention. HSS T3 HPLC columns are fully scalable and transferable to UPLC separations.
  • Trifunctional alkyl C18 bonded phase, compatible with 100% aqueous mobile phase
  • Provides ultra-low MS bleed
  • Promotes superior polar compound retention
  • Fully scalable and transferable to UPLC separations
Catalog No. 50-179-712 Supplier Waters Corp Supplier No. 186006458
The trifunctional alkyl C18 bonded phase used for the HSS T3 sorbents is compatible with 100% aqueous mobile phase and provides ultra-low MS bleed, while promoting superior polar compound retention. HSS T3 HPLC columns are fully scalable and transferable to UPLC separations.
  • Trifunctional alkyl C18 bonded phase, compatible with 100% aqueous mobile phase
  • Provides ultra-low MS bleed
  • Promotes superior polar compound retention
  • Fully scalable and transferable to UPLC separations