Learn More
Waters Corp Waters XSelect HSS T3 Column, 100Å, 3.5 μm, 2.1 mm X 30 mm
Catalog No. 50179716 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The trifunctional alkyl C18 bonded phase is compatible with 100% aqueous mobile phase and provides ultra-low MS bleed, while promoting superior polar compound retention. These HPLC columns are fully scalable and transferable to UPLC separations.
  • Trifunctional alkyl C18 bonded phase compatible with 100% aqueous mobile phase
  • Provides ultra-low MS bleed
  • Promotes superior polar compound retention
  • Fully scalable and transferable to UPLC separations
Catalog No. 50-179-716 Supplier Waters Corp Supplier No. 186006462
Add to Cart
Add to Cart
The trifunctional alkyl C18 bonded phase is compatible with 100% aqueous mobile phase and provides ultra-low MS bleed, while promoting superior polar compound retention. These HPLC columns are fully scalable and transferable to UPLC separations.
  • Trifunctional alkyl C18 bonded phase compatible with 100% aqueous mobile phase
  • Provides ultra-low MS bleed
  • Promotes superior polar compound retention
  • Fully scalable and transferable to UPLC separations