Learn More
Waters Corp Waters Column, C18, 2.5 μm, 4.6 mm, 150 mm
Supplier:  Waters Corp 186006741
Encompass_Preferred
This column is a low-coverage C18 phase designed for reversed-phase separations of both polar and non-polar compounds, compatible with 100% aqueous mobile phases. It delivers exceptional separation performance, robustness, and throughput for HPLC assays, providing a seamless transition to UPLC.
  • Compatible with 100% aqueous mobile phase.
  • Ultra-low MS bleed.
  • Scalable and transferable to UPLC separations.
  • Achieves higher retention for polar compounds.
  • Enhanced selectivity for acidic, alkaline, and neutral compounds.
  • Built on HSS ‘High Strength Silica’ Technology for mechanical strength and increased polar compound retention.
  • Outstanding reproducibility and transferability.
  • Recommended operational pH range between 2 and 8.
Catalog No. 50-466-449