Learn More
Waters Corp XSelect HSS T3 Column, 5 μm, 100 Å, 4.6 mm X 75 mm
Catalog No. 50179738 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns utilize High Strength Silica (HSS) Technology for excellent peak shape, high efficiency, and superior reversed-phase retention of polar analytes, ensuring robust performance and seamless method transfer across chromatography platforms.
  • Manufactured in a cGMP, ISO 9001 certified plant with ultra-pure reagents.
  • Batches are chromatographically tested for consistent, reproducible performance.
  • Each column is individually tested; includes a performance chromatogram and certificate of analysis.
  • Documents accessible via eCord intelligent chip, eConnect tag, or QR code.
  • Designed for superior reversed-phase retention of polar analytes.
  • Optimal operation between 20°C to 80°C enhances selectivity and mass transfer.
  • Features 100 Å pore diameter and 230 m²/g surface area.
  • Maintains performance within a pH range of 2–8.
Catalog No. 50-179-738 Supplier Waters Corp Supplier No. 186006484
These columns utilize High Strength Silica (HSS) Technology for excellent peak shape, high efficiency, and superior reversed-phase retention of polar analytes, ensuring robust performance and seamless method transfer across chromatography platforms.
  • Manufactured in a cGMP, ISO 9001 certified plant with ultra-pure reagents.
  • Batches are chromatographically tested for consistent, reproducible performance.
  • Each column is individually tested; includes a performance chromatogram and certificate of analysis.
  • Documents accessible via eCord intelligent chip, eConnect tag, or QR code.
  • Designed for superior reversed-phase retention of polar analytes.
  • Optimal operation between 20°C to 80°C enhances selectivity and mass transfer.
  • Features 100 Å pore diameter and 230 m²/g surface area.
  • Maintains performance within a pH range of 2–8.