Learn More
Waters Corp XSelect HSS T3 Column, 100Å, 5 μm, 4.6 mm X 250 mm
Supplier:  Waters Corp 186004793RF
Encompass_Preferred
The HSS T3 column is a fully endcapped, low-coverage C18 bonded-phase column designed to retain extremely polar compounds by reversed-phase in high aqueous mobile phases. It utilizes high strength silica technology, offering higher silanophilicity for increased polar compound retention and significantly different selectivity. This technology also provides the mechanical strength for UPLC pressures, supreme reproducibility, and seamless scalability. It facilitates higher resolution separations for complex samples, combining efficiency and selectivity.
  • Fully endcapped, low-coverage C18 bonded-phase for specific retention.
  • Designed to retain extremely polar compounds by reversed-phase in high aqueous mobile phases.
  • Utilizes high strength silica technology.
  • Possesses mechanical strength necessary for UPLC pressures.
Catalog No. 50-239-1051