Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Column, C18, 100 Å, 5 μm, 2.1 mm x 250 mm

Catalog No. 502391534 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

XSelect Peptide HSS T3 Columns are designed for silica-based separations requiring increased retention of hydrophilic peptides. Each batch of HSS T3, 100Å material is QC tested with a gradient separation of a tryptic digest of cytochrome c, ensuring stringent performance.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Silica
  • PH range minimum: 2 pH
  • PH range maximum: 8 pH
  • Temperature limits: 45°C
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Bonding technology: T3
  • Silanol activity: Medium
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Pore size: 100 Å
  • Inner diameter: 2.1 mm
  • Length: 250 mm
  • Carbon load: 11%

Catalog No. 50-239-1534 Supplier Waters Corp Supplier No. 186008869
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XSelect Peptide HSS T3 Columns are designed for silica-based separations requiring increased retention of hydrophilic peptides. Each batch of HSS T3, 100Å material is QC tested with a gradient separation of a tryptic digest of cytochrome c, ensuring stringent performance.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Silica
  • PH range minimum: 2 pH
  • PH range maximum: 8 pH
  • Temperature limits: 45°C
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Bonding technology: T3
  • Silanol activity: Medium
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Pore size: 100 Å
  • Inner diameter: 2.1 mm
  • Length: 250 mm
  • Carbon load: 11%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.