Learn More
Waters Corp Waters Method Validation Kit, XSelect HSS T3, 3.5 μm
Catalog No. 50201637 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The trifunctional alkyl C18 bonded phase used for the HSS T3 sorbents is compatible with 100% aqueous mobile phase and provides ultra-low MS bleed, while promoting superior polar compound retention. HSS T3 HPLC columns are fully scalable and transferable to UPLC separations.
  • Compatible with 100% aqueous mobile phase.
  • Provides ultra-low MS bleed.
  • Promotes superior polar compound retention.
  • Fully scalable and transferable to UPLC separations.
Catalog No. 50-201-637 Supplier Waters Corp Supplier No. 186006492
The trifunctional alkyl C18 bonded phase used for the HSS T3 sorbents is compatible with 100% aqueous mobile phase and provides ultra-low MS bleed, while promoting superior polar compound retention. HSS T3 HPLC columns are fully scalable and transferable to UPLC separations.
  • Compatible with 100% aqueous mobile phase.
  • Provides ultra-low MS bleed.
  • Promotes superior polar compound retention.
  • Fully scalable and transferable to UPLC separations.