Learn More
Waters Corp Waters XSelect HSS T3 Kit, 5 μm, 4.6 x 100 mm
Supplier:  Waters Corp 186006496
Encompass_Preferred
The XSelect HSS T3 Method Validation Kit is designed for reversed-phase applications, providing increased retention for polar compounds and broad retention capabilities for mixtures of polar and non-polar compounds. The columns utilize High Strength Silica Technology engineered to withstand UPLC pressures.
  • Increased retention for polar compounds compared to standard C18 phases.
  • Suitable for reversed-phase applications, even with up to 100% aqueous mobile phases.
  • Broad retention capabilities for mixtures of polar and non-polar compounds with standard reversed-phase mobile phases.
  • High Strength Silica Technology columns engineered to withstand UPLC pressures.
  • Kit includes three columns, each from a distinct batch.
Catalog No. 50-201-641