Learn More
Waters Corp Waters XSelect HSS T3 Method Validation Kit, C18, 3.5 μm
Catalog No. 50201636 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The HSS T3 Method Validation Kit utilizes a trifunctional alkyl C18 bonded phase compatible with 100% aqueous mobile phase, offering ultra-low MS bleed and enhancing polar compound retention. These HPLC columns are scalable and transferable to UPLC separations, designed to improve selectivity for acidic, alkaline, and neutral chemicals, and enhance the resolution of complex samples.
  • Compatible with 100% aqueous mobile phase and provides ultra-low MS bleed.
  • Enhances polar compound retention and improves selectivity for various chemicals.
  • Offers exceptional repeatability and transferability between HPLC and UPLC systems.
  • Manufactured in a cGMP, ISO 9001 certified facility to high industrial standards.
  • Provides wider retention capabilities for combinations of polar and non-polar chemicals.
Catalog No. 50-201-636 Supplier Waters Corp Supplier No. 186006491
The HSS T3 Method Validation Kit utilizes a trifunctional alkyl C18 bonded phase compatible with 100% aqueous mobile phase, offering ultra-low MS bleed and enhancing polar compound retention. These HPLC columns are scalable and transferable to UPLC separations, designed to improve selectivity for acidic, alkaline, and neutral chemicals, and enhance the resolution of complex samples.
  • Compatible with 100% aqueous mobile phase and provides ultra-low MS bleed.
  • Enhances polar compound retention and improves selectivity for various chemicals.
  • Offers exceptional repeatability and transferability between HPLC and UPLC systems.
  • Manufactured in a cGMP, ISO 9001 certified facility to high industrial standards.
  • Provides wider retention capabilities for combinations of polar and non-polar chemicals.