Learn More
Waters Corp Waters XSelect HSS T3 Method Validation Kit, 100Å, 3.5 μm, 3 mm × 150 mm, 3/pk
Catalog No. 50201635 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The trifunctional alkyl C18 bonded phase used for the HSS T3 sorbents is compatible with 100% aqueous mobile phase and provides ultra-low MS bleed, while promoting superior polar compound retention. HSS T3 HPLC columns are fully scalable and transferable to UPLC separations.
  • C18 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Temperature limits up to 45°C
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • T3 bonding technology
  • Medium silanol activity
  • Spherical particle shape
  • 3.5 μm particle size
  • 100 Å pore size
  • Method validation kit format
  • 230 surface area
  • HPLC system compatible
  • HSS particle technology
  • L1 USP classification
  • 3 mm inner diameter
  • 150 mm length
  • 11% carbon load
  • 3 units per package
Catalog No. 50-201-635 Supplier Waters Corp Supplier No. 186006490
The trifunctional alkyl C18 bonded phase used for the HSS T3 sorbents is compatible with 100% aqueous mobile phase and provides ultra-low MS bleed, while promoting superior polar compound retention. HSS T3 HPLC columns are fully scalable and transferable to UPLC separations.
  • C18 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Temperature limits up to 45°C
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • T3 bonding technology
  • Medium silanol activity
  • Spherical particle shape
  • 3.5 μm particle size
  • 100 Å pore size
  • Method validation kit format
  • 230 surface area
  • HPLC system compatible
  • HSS particle technology
  • L1 USP classification
  • 3 mm inner diameter
  • 150 mm length
  • 11% carbon load
  • 3 units per package