Learn More
Waters Corp Waters Column Kit, Method Validation, 100Å, 3.5 μm, 3 mm X 100 mm
Supplier:  Waters Corp 186006489
Encompass_Preferred
The trifunctional alkyl C18 bonded phase utilized in these HSS T3 sorbents ensures compatibility with 100% aqueous mobile phases and delivers ultra-low MS bleed. It also enhances retention of polar compounds, making these HPLC columns fully scalable and transferable to UPLC separations.
  • Trifunctional alkyl C18 bonded phase.
  • Compatible with 100% aqueous mobile phase.
  • Provides ultra-low MS bleed.
  • Promotes superior polar compound retention.
  • Fully scalable and transferable to UPLC separations.
Catalog No. 50-201-634