Learn More
Waters Corp XSelect HSS T3 Method Validation Kit, 100Å, 5 µm, 4.6 mm X 150 mm, 3/pk
Catalog No. 50201642 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The trifunctional alkyl C18 bonded phase used for the HSS T3 sorbents is compatible with 100% aqueous mobile phase and provides ultra-low MS bleed, while promoting superior polar compound retention. HSS T3 HPLC columns are fully scalable and transferable to UPLC separations.
Catalog No. 50-201-642 Supplier Waters Corp Supplier No. 186006497
The trifunctional alkyl C18 bonded phase used for the HSS T3 sorbents is compatible with 100% aqueous mobile phase and provides ultra-low MS bleed, while promoting superior polar compound retention. HSS T3 HPLC columns are fully scalable and transferable to UPLC separations.