Learn More
Waters Corp VanGuard Cartridge, 100Å, 3.5 μm, 2.1 mm x 5 mm, 3/pk
Catalog No. 502391073 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect HSS T3 VanGuard Cartridges extend the lifetime and performance of analytical columns by removing particulate contamination. This cartridge is optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns with 3.5 μm sorbent particles. Practical, easy-to-use, and disposable, they offer a cost-effective solution to safeguard more expensive columns. They provide twice the capacity of conventional guard columns, reducing column changes and improving repeatability.
  • Extends analytical column lifetime and performance
  • Removes particulate contamination from the mobile phase stream
  • Optimized for 2.1 mm and 3.0 mm I.D. analytical columns with 3.5 μm sorbent particles
  • Contains high-performance packing materials
  • Disposable and cost-effective
  • Quick and easy to change
  • Offers twice the capacity of conventional guard columns
  • Improves repeatability of results
Catalog No. 50-239-1073 Supplier Waters Corp Supplier No. 186007878
XSelect HSS T3 VanGuard Cartridges extend the lifetime and performance of analytical columns by removing particulate contamination. This cartridge is optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns with 3.5 μm sorbent particles. Practical, easy-to-use, and disposable, they offer a cost-effective solution to safeguard more expensive columns. They provide twice the capacity of conventional guard columns, reducing column changes and improving repeatability.
  • Extends analytical column lifetime and performance
  • Removes particulate contamination from the mobile phase stream
  • Optimized for 2.1 mm and 3.0 mm I.D. analytical columns with 3.5 μm sorbent particles
  • Contains high-performance packing materials
  • Disposable and cost-effective
  • Quick and easy to change
  • Offers twice the capacity of conventional guard columns
  • Improves repeatability of results