Learn More
Waters Corp VanGuard Cartridge, 100Å, 5 μm, 3.9 mm × 5 mm, 3 pk
Catalog No. 502391256 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect HSS T3 VanGuard Cartridges are used to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 4.6 mm I.D. XSelect HSS T3 analytical columns containing 5 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect 4.6 mm I.D. analytical columns containing 5 μm sorbent particles.
Catalog No. 50-239-1256 Supplier Waters Corp Supplier No. 186007883
XSelect HSS T3 VanGuard Cartridges are used to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 4.6 mm I.D. XSelect HSS T3 analytical columns containing 5 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect 4.6 mm I.D. analytical columns containing 5 μm sorbent particles.