Learn More
Waters Corp Waters Corporation Column, C18, 100Å 2.5 μm, 4.6 mm × 100 mm
Catalog No. 50201564 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column features a low-coverage C18 phase compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. It exhibits ultra-low MS bleed, offers full scalability and transferability to UPLC separations. The 2.5 μm eXtended Performance (XP) columns, packed in ultra-low dispersion hardware, deliver exceptional HPLC performance, robustness, throughput, and facilitate UPLC transition.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phase.
  • Exhibits ultra-low MS bleed.
  • Fully scalable and transferable to UPLC separations.
  • Provides exceptional separation performance, robustness, and throughput for HPLC assays.
  • Offers maximum transferability and reproducibility for higher resolution separations.
  • High Strength Silica Technology (HSS Technology) boosts polar retention and selectivity.
Catalog No. 50-201-564 Supplier Waters Corp Supplier No. 186006159
This column features a low-coverage C18 phase compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. It exhibits ultra-low MS bleed, offers full scalability and transferability to UPLC separations. The 2.5 μm eXtended Performance (XP) columns, packed in ultra-low dispersion hardware, deliver exceptional HPLC performance, robustness, throughput, and facilitate UPLC transition.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phase.
  • Exhibits ultra-low MS bleed.
  • Fully scalable and transferable to UPLC separations.
  • Provides exceptional separation performance, robustness, and throughput for HPLC assays.
  • Offers maximum transferability and reproducibility for higher resolution separations.
  • High Strength Silica Technology (HSS Technology) boosts polar retention and selectivity.