Learn More
Waters Corp Waters XSelect HSS T3 Column, 100Å, 2.5 μm, 2.1 mm × 30 mm
Supplier:  Waters Corp 186006148
Encompass_Preferred
The XSelect HSS T3 column features a low-coverage C18 phase, ensuring compatibility with 100% aqueous mobile phases for separating both polar and non-polar compounds. These columns are designed for ultra-low MS bleed and offer scalability for UPLC separations.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phase.
  • Ideal for developing separations of both polar and non-polar compounds.
  • Characterized by ultra-low MS bleed.
  • Fully scalable and transferable to UPLC separations.
  • 2.5 μm eXtended Performance [XP] columns packed in ultra-low dispersion hardware.
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.
  • Provides a seamless transition path toward UPLC adoption.
  • All column documents, including Certificates of Analysis (COAs), are available at www.waters.com/coa for easy access.
Catalog No. 50-179-544