Learn More
Waters Corp Waters Column, C18, 2.5 μm, 3 mm X 30 mm, 3 pk
Catalog No. 50179289 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS T3 is a low-coverage C18 phase column, compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. These columns feature ultra-low MS bleed and are scalable to UPLC separations. The 2.5 μm eXtended Performance (XP) columns offer exceptional separation, robustness, and throughput for HPLC, with a clear path to UPLC.
  • Low-coverage C18 phase.
  • Compatible with 100% aqueous mobile phase.
  • Suitable for separating polar and non-polar compounds.
  • Exhibits ultra-low MS bleed.
  • Fully scalable and transferable to UPLC separations.
  • Packed in ultra-low dispersion hardware for eXtended Performance (XP).
  • Enables exceptional separation performance, robustness, and throughput for HPLC.
  • Provides a seamless transition path toward UPLC adoption.
Catalog No. 50-179-289 Supplier Waters Corp Supplier No. 176002630
The XSelect HSS T3 is a low-coverage C18 phase column, compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. These columns feature ultra-low MS bleed and are scalable to UPLC separations. The 2.5 μm eXtended Performance (XP) columns offer exceptional separation, robustness, and throughput for HPLC, with a clear path to UPLC.
  • Low-coverage C18 phase.
  • Compatible with 100% aqueous mobile phase.
  • Suitable for separating polar and non-polar compounds.
  • Exhibits ultra-low MS bleed.
  • Fully scalable and transferable to UPLC separations.
  • Packed in ultra-low dispersion hardware for eXtended Performance (XP).
  • Enables exceptional separation performance, robustness, and throughput for HPLC.
  • Provides a seamless transition path toward UPLC adoption.