Learn More
Waters Corp Chromatography Column, XSelect HSS T3, 2.5 μm, 50 mm, 3.0 mm ID
Catalog No. 50179618 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS T3 XP Column is a chromatography column designed to bridge the gap between HPLC and UPLC™ Technology, offering enhanced performance and flexibility for various LC systems. This column provides exceptional separation performance and robustness, making it suitable for a wide range of laboratory applications.
  • Compatible with HPLC, UHPLC, and UPLC systems
  • Improves productivity with intermediate backpressures, offering 40% lower backpressure than 1.7 μm UPLC columns
  • Directly scalable to HPLC and UPLC columns for easy method transfer
  • Provides exceptional efficiency and mass transfer for robust HPLC assays
  • Enables rapid method optimization
  • Demonstrates superior loading capacity for complex mixtures compared to equivalent core-shell particle columns
  • Helps future-proof laboratories with UPLC Technology
Catalog No. 50-179-618 Supplier Waters Corp Supplier No. 186006259
The XSelect HSS T3 XP Column is a chromatography column designed to bridge the gap between HPLC and UPLC™ Technology, offering enhanced performance and flexibility for various LC systems. This column provides exceptional separation performance and robustness, making it suitable for a wide range of laboratory applications.
  • Compatible with HPLC, UHPLC, and UPLC systems
  • Improves productivity with intermediate backpressures, offering 40% lower backpressure than 1.7 μm UPLC columns
  • Directly scalable to HPLC and UPLC columns for easy method transfer
  • Provides exceptional efficiency and mass transfer for robust HPLC assays
  • Enables rapid method optimization
  • Demonstrates superior loading capacity for complex mixtures compared to equivalent core-shell particle columns
  • Helps future-proof laboratories with UPLC Technology