Learn More
Waters Corp XSelect HSS T3 XP Column, 100 Å, 2.5 μm, 3 mm x 50 mm
Catalog No. 50179290 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS T3 XP Column is a low-coverage C18 phase for separating polar and non-polar compounds, compatible with 100% aqueous mobile phases. Its 2.5 μm eXtended Performance [XP] columns, featuring ultra-low dispersion hardware, deliver exceptional HPLC performance and provide a seamless path to UPLC.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phases for polar and non-polar compound separations.
  • Ultra-low MS bleed and full scalability to UPLC separations.
  • 2.5 μm XP columns with ultra-low dispersion hardware for exceptional HPLC performance.
  • Wide operating range: pH 2 to 8, up to 45 °C, and pressures up to 18000 psi (1240 Bar).
  • Increased polar compound retention and broad retention capability.
  • Manufactured to cGMP, ISO 9001 standards with individual testing.
Catalog No. 50-179-290 Supplier Waters Corp Supplier No. 176002631
The XSelect HSS T3 XP Column is a low-coverage C18 phase for separating polar and non-polar compounds, compatible with 100% aqueous mobile phases. Its 2.5 μm eXtended Performance [XP] columns, featuring ultra-low dispersion hardware, deliver exceptional HPLC performance and provide a seamless path to UPLC.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phases for polar and non-polar compound separations.
  • Ultra-low MS bleed and full scalability to UPLC separations.
  • 2.5 μm XP columns with ultra-low dispersion hardware for exceptional HPLC performance.
  • Wide operating range: pH 2 to 8, up to 45 °C, and pressures up to 18000 psi (1240 Bar).
  • Increased polar compound retention and broad retention capability.
  • Manufactured to cGMP, ISO 9001 standards with individual testing.