Learn More
Waters Corp Waters XSelect HSS T3 Column, 100Å, 2.5 μm, 2.1 mm X 50 mm
Catalog No. 50179545 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS T3 XP Column is a low-coverage C18 phase compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. It offers ultra-low MS bleed and scalability to UPLC. Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance [XP] columns provide exceptional separation performance, robustness, and throughput for HPLC assays, with a seamless transition to UPLC.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phase
  • Separates polar and non-polar compounds; ultra-low MS bleed
  • Scalable and transferable to UPLC; exceptional HPLC performance and robustness
  • Increased retention for polar compounds; widest retention for mixed analytes; enhanced selectivity
  • Manufactured to cGMP, ISO 9001 standards; individually tested for quality
Catalog No. 50-179-545 Supplier Waters Corp Supplier No. 186006149
The XSelect HSS T3 XP Column is a low-coverage C18 phase compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. It offers ultra-low MS bleed and scalability to UPLC. Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance [XP] columns provide exceptional separation performance, robustness, and throughput for HPLC assays, with a seamless transition to UPLC.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phase
  • Separates polar and non-polar compounds; ultra-low MS bleed
  • Scalable and transferable to UPLC; exceptional HPLC performance and robustness
  • Increased retention for polar compounds; widest retention for mixed analytes; enhanced selectivity
  • Manufactured to cGMP, ISO 9001 standards; individually tested for quality