Learn More
Waters Corp Waters XSelect HSS T3 XP Column, C18 Chemistry, 2.5 μm Particle Size, 100Å Pore, 3 mm x 100 mm
Supplier: Waters Corp 186006155
This column features a low-coverage C18 phase, ideal for separating both polar and non-polar compounds, and is fully compatible with 100% aqueous mobile phases. It ensures ultra-low MS bleed and provides exceptional separation performance and robustness for HPLC assays. The column is fully scalable and transferable, enabling a smooth transition to UPLC separations and adoption.
- C18 chemistry and reversed-phase separation mode
- Compatible with 100% aqueous mobile phases
- Offers ultra-low MS bleed
- Features 2.5 μm particle size and 100 Å pore size
- Designed for high pressure applications up to 18000 psi (1240 Bar)
- Broad operating pH range of 2 to 8
- Provides exceptional separation performance and robustness
- Facilitates a seamless transition to UPLC adoption
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.