Learn More
Waters Corp Waters Corporation Column, XSelect HSS T3, 100Å, 2.5 μm
Catalog No. 50201562 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS T3 Column features a trifunctional alkyl C18 bonded phase, offering compatibility with 100% aqueous mobile phase, ultra-low MS bleed, and enhanced polar compound retention. Utilizing HSS (High Strength Silica) technology, it provides improved chromatographic performance and the mechanical strength required for UPLC pressures, making it fully scalable and transferable to UPLC separations.
  • Compatible with 100% aqueous mobile phase and offers ultra-low MS bleed.
  • Enhances retention of polar compounds.
  • Fully scalable and transferable to UPLC separations.
  • HSS (High Strength Silica) technology improves chromatographic performance and provides mechanical strength for UPLC pressures.
  • Designed to retain highly polar chemicals in high-aqueous mobile phases.
  • Enables better resolution for challenging and complex samples.
Catalog No. 50-201-562 Supplier Waters Corp Supplier No. 186006157
Add to Cart
Add to Cart
The XSelect HSS T3 Column features a trifunctional alkyl C18 bonded phase, offering compatibility with 100% aqueous mobile phase, ultra-low MS bleed, and enhanced polar compound retention. Utilizing HSS (High Strength Silica) technology, it provides improved chromatographic performance and the mechanical strength required for UPLC pressures, making it fully scalable and transferable to UPLC separations.
  • Compatible with 100% aqueous mobile phase and offers ultra-low MS bleed.
  • Enhances retention of polar compounds.
  • Fully scalable and transferable to UPLC separations.
  • HSS (High Strength Silica) technology improves chromatographic performance and provides mechanical strength for UPLC pressures.
  • Designed to retain highly polar chemicals in high-aqueous mobile phases.
  • Enables better resolution for challenging and complex samples.