Learn More
Waters Corp Waters XSelect HSS T3 Column, 100Å, 2.5 μm, 2.1 mm x 100 mm
Catalog No. 50179547 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS T3 column is a low-coverage C18 phase, compatible with 100% aqueous mobile phases. It effectively separates polar and non-polar compounds, offering exceptional performance, robustness, and high throughput for HPLC assays and seamless transition to UPLC.
  • Low-coverage C18 phase, compatible with 100% aqueous mobile phases.
  • Effectively separates polar and non-polar compounds.
  • Exceptional separation performance, robustness, and high throughput for HPLC.
  • Seamless scalability and transferability to UPLC applications.
  • Increased retention for polar compounds vs. traditional C18 phases.
  • Enhanced selectivity for diverse compounds, improving complex sample resolution.
  • Manufactured under cGMP, ISO 9001, and individually tested for reliable results.
  • Withstands pressures up to 6000 psi (400 bar or 40 MPa).
Catalog No. 50-179-547 Supplier Waters Corp Supplier No. 186006151
The XSelect HSS T3 column is a low-coverage C18 phase, compatible with 100% aqueous mobile phases. It effectively separates polar and non-polar compounds, offering exceptional performance, robustness, and high throughput for HPLC assays and seamless transition to UPLC.
  • Low-coverage C18 phase, compatible with 100% aqueous mobile phases.
  • Effectively separates polar and non-polar compounds.
  • Exceptional separation performance, robustness, and high throughput for HPLC.
  • Seamless scalability and transferability to UPLC applications.
  • Increased retention for polar compounds vs. traditional C18 phases.
  • Enhanced selectivity for diverse compounds, improving complex sample resolution.
  • Manufactured under cGMP, ISO 9001, and individually tested for reliable results.
  • Withstands pressures up to 6000 psi (400 bar or 40 MPa).