Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Column, C18, 100Å, 2.5 μm, 2.1 mm, 100 mm

Catalog No. 50179288 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XSelect HSS T3 Column features a low-coverage C18 phase, ideal for separating polar and non-polar compounds with 100% aqueous mobile phase. These 2.5 μm XP columns provide robust, high-performance separation for HPLC assays, offering a seamless transition to UPLC.

  • Low-coverage C18 phase for polar and non-polar compounds.
  • Ultra-low MS bleed.
  • HSS (High Strength Silica) technology provides mechanical strength.
  • Increased polar compound retention and different selectivity.
  • Pore size: 100 Å.
  • Particle size: 2.5 μm.
  • Inner diameter: 2.1 mm.
  • Length: 100 mm.
  • Maximum pressure: 18000 psi (1240 Bar).
  • pH range: 2 to 8 pH.
  • Units per package: 3 pk.

Catalog No. 50-179-288 Supplier Waters Corp Supplier No. 176002629
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XSelect HSS T3 Column features a low-coverage C18 phase, ideal for separating polar and non-polar compounds with 100% aqueous mobile phase. These 2.5 μm XP columns provide robust, high-performance separation for HPLC assays, offering a seamless transition to UPLC.

  • Low-coverage C18 phase for polar and non-polar compounds.
  • Ultra-low MS bleed.
  • HSS (High Strength Silica) technology provides mechanical strength.
  • Increased polar compound retention and different selectivity.
  • Pore size: 100 Å.
  • Particle size: 2.5 μm.
  • Inner diameter: 2.1 mm.
  • Length: 100 mm.
  • Maximum pressure: 18000 psi (1240 Bar).
  • pH range: 2 to 8 pH.
  • Units per package: 3 pk.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.