Learn More
Waters Corp Waters XSelect HSS T3 Column, 100Å, 2.5 μm, 2.1 mm × 75 mm
Catalog No. 50179546 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS T3 is a low-coverage C18 phase compatible with 100% aqueous mobile phase, designed for separations of polar and non-polar compounds. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, offering exceptional performance for HPLC assays and a seamless transition to UPLC.
  • Compatible with 100% aqueous mobile phase
  • Ultra-low MS bleed
  • Scalable and transferable to UPLC separations
  • Improved selectivity for acidic, alkaline, and neutral chemicals
  • Enhanced polar component retention
  • Exceptional repeatability and transferability
  • Higher retention for polar chemicals than standard C18 phase
  • Manufactured in a cGMP, ISO 9001 certified facility
  • Columns tested to ensure high-quality, consistent results
  • Includes an eCord™ Intelligent Chip with performance chromatogram and certificate of batch analysis
Catalog No. 50-179-546 Supplier Waters Corp Supplier No. 186006150
The XSelect HSS T3 is a low-coverage C18 phase compatible with 100% aqueous mobile phase, designed for separations of polar and non-polar compounds. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, offering exceptional performance for HPLC assays and a seamless transition to UPLC.
  • Compatible with 100% aqueous mobile phase
  • Ultra-low MS bleed
  • Scalable and transferable to UPLC separations
  • Improved selectivity for acidic, alkaline, and neutral chemicals
  • Enhanced polar component retention
  • Exceptional repeatability and transferability
  • Higher retention for polar chemicals than standard C18 phase
  • Manufactured in a cGMP, ISO 9001 certified facility
  • Columns tested to ensure high-quality, consistent results
  • Includes an eCord™ Intelligent Chip with performance chromatogram and certificate of batch analysis