Learn More
Waters Corp Waters XSelect HSS T3 Column, 100Å, 2.5 μm, 3 mm x 100 mm
Supplier:  Waters Corp 176002633
Encompass_Preferred
This column features a low-coverage C18 phase compatible with 100% aqueous mobile phase, designed for separating polar and non-polar compounds with ultra-low MS bleed. It offers exceptional performance for HPLC assays and a transition path to UPLC.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phase
  • Designed for developing separations for polar and non-polar compounds
  • Exhibits ultra-low MS bleed
  • Fully scalable and transferable to UPLC separations
  • Packed in ultra-low dispersion hardware for eXtended performance (XP)
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays
  • Provides a seamless transition path toward UPLC adoption
Catalog No. 50-179-292