Learn More
Waters Corp Waters Column, XSelect HSS T3, 100Å, 2.5 μm, 3 mm ID x 75 mm
Catalog No. 50179291 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS T3 column features a low-coverage C18 phase, compatible with 100% aqueous mobile phases. It enables effective separation of polar and non-polar compounds, offering ultra-low MS bleed and UPLC scalability. Designed for enhanced selectivity, it improves resolution for various chemicals and simplifies chromatographic development.
  • Low-coverage C18 phase
  • 100% aqueous mobile phase compatible
  • Ultra-low MS bleed; UPLC scalable
  • Enhanced selectivity for diverse chemicals
  • Exceptional HPLC performance (2.5 μm XP hardware)
  • Superior polar component retention (high silanophilicity)
  • Reproducible, manufactured to cGMP, ISO 9001 standards
Catalog No. 50-179-291 Supplier Waters Corp Supplier No. 176002632
The XSelect HSS T3 column features a low-coverage C18 phase, compatible with 100% aqueous mobile phases. It enables effective separation of polar and non-polar compounds, offering ultra-low MS bleed and UPLC scalability. Designed for enhanced selectivity, it improves resolution for various chemicals and simplifies chromatographic development.
  • Low-coverage C18 phase
  • 100% aqueous mobile phase compatible
  • Ultra-low MS bleed; UPLC scalable
  • Enhanced selectivity for diverse chemicals
  • Exceptional HPLC performance (2.5 μm XP hardware)
  • Superior polar component retention (high silanophilicity)
  • Reproducible, manufactured to cGMP, ISO 9001 standards