Learn More
Waters Corp Waters XSelect HSS T3 Column, 2.5 μm, 2.1 mm ID, 50 mm Length, 100Å
Catalog No. 50179286 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS T3 column features a low-coverage C18 phase compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. It exhibits ultra-low MS bleed, offering full scalability and transferability to UPLC separations. These 2.5 μm eXtended Performance (XP) columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a seamless transition to UPLC adoption.
  • Low-coverage C18 phase for polar and non-polar separations in 100% aqueous mobile phase.
  • Ultra-low MS bleed, fully scalable and transferable to UPLC methods.
  • eXtended Performance (XP) columns provide exceptional HPLC separation performance and robustness.
  • HSS technology ensures improved chromatography, strong retention of polar chemicals, and withstands UPLC pressures.
Catalog No. 50-179-286 Supplier Waters Corp Supplier No. 176002627
The XSelect HSS T3 column features a low-coverage C18 phase compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. It exhibits ultra-low MS bleed, offering full scalability and transferability to UPLC separations. These 2.5 μm eXtended Performance (XP) columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a seamless transition to UPLC adoption.
  • Low-coverage C18 phase for polar and non-polar separations in 100% aqueous mobile phase.
  • Ultra-low MS bleed, fully scalable and transferable to UPLC methods.
  • eXtended Performance (XP) columns provide exceptional HPLC separation performance and robustness.
  • HSS technology ensures improved chromatography, strong retention of polar chemicals, and withstands UPLC pressures.