Learn More
Waters Corp XP VanGuard Cartridge, 100Å, 2.5 μm, 2.1 mm X 5 mm, 3 pk
Catalog No. 502390535 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
VanGuard Cartridges extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase. Optimized to protect 2.1 mm and 3.0 mm I.D. HSS T3 analytical columns with 2.5 μm sorbent particles. Designed for UPLC columns in reversed-phase liquid chromatography, it helps maintain separation efficiency in UPLC, UHPLC, and HPLC by removing chemical and particle contaminants.
  • C18 chemistry and reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Temperature limits of 45°C
  • Maximum pressure of 18000 psi (1240 bar)
  • T3 bonding and HSS particle technology
  • 2.5 μm particle size
  • 100Å pore size
  • 2.1 mm inner diameter and 5 mm length
  • Compatible with UPLC and UHPLC systems
  • 3 pk units per package
Catalog No. 50-239-0535 Supplier Waters Corp Supplier No. 186007884
VanGuard Cartridges extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase. Optimized to protect 2.1 mm and 3.0 mm I.D. HSS T3 analytical columns with 2.5 μm sorbent particles. Designed for UPLC columns in reversed-phase liquid chromatography, it helps maintain separation efficiency in UPLC, UHPLC, and HPLC by removing chemical and particle contaminants.
  • C18 chemistry and reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Temperature limits of 45°C
  • Maximum pressure of 18000 psi (1240 bar)
  • T3 bonding and HSS particle technology
  • 2.5 μm particle size
  • 100Å pore size
  • 2.1 mm inner diameter and 5 mm length
  • Compatible with UPLC and UHPLC systems
  • 3 pk units per package