Learn More
Waters Corp XSelect Peptide CSH C18 Column, 130Å, 3.5 μm, 4.6 mm × 150 mm
Supplier:  Waters Corp 186006957
Encompass_Preferred
This analytical column features Charged Surface Hybrid (CSH) technology particles, providing superior peak form and enhanced basic compound loading capacity. It delivers excellent peak shape for peptides using either FA- or TFA-containing mobile phases, ideal for LC or LC-MS applications.
  • Superior peak form and basic compound loading capacity with CSH technology.
  • Higher peptide mass loading for enhanced detection of low-level constituents.
  • Remarkable peak capabilities for peptides and small proteins up to 10,000 Daltons in HPLC, UPLC, or LC-MS.
  • Scalable chemistries from nanoflow analysis to lab-scale purification.
  • Flexible eluent choice (TFA or FA) due to low reliance on strong ion pairing agents.
Catalog No. 50-239-2392