Learn More
Waters Corp XSelect Peptide CSH C18 Column Kit, 130Å, 2.5 μm, 2.1 mm X 50 mm
Catalog No. 502392142 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These column kits offer excellent peak shape for peptides in FA- or TFA-containing mobile phases, ideal for LC or LC-MS applications due to minimal dependence on strong ion pairing agents. With ultra-low dispersion hardware, they ensure exceptional separation, robustness, and high throughput for HPLC, enabling a smooth transition to UPLC.
  • Low, carefully-defined positive charge for excellent peptide peak shape.
  • Minimal dependence on strong ion pairing agents, ideal for LC or LC-MS.
  • Ultra-low dispersion hardware for exceptional separation, robustness, and high throughput in HPLC.
  • Seamless transition path toward UPLC adoption.
  • Includes column and Cytochrome C Digestion Standard for immediate benchmarking.
  • Digestion standard batches under strict quality control with certificate of analysis.
Catalog No. 50-239-2142 Supplier Waters Corp Supplier No. 176003069
These column kits offer excellent peak shape for peptides in FA- or TFA-containing mobile phases, ideal for LC or LC-MS applications due to minimal dependence on strong ion pairing agents. With ultra-low dispersion hardware, they ensure exceptional separation, robustness, and high throughput for HPLC, enabling a smooth transition to UPLC.
  • Low, carefully-defined positive charge for excellent peptide peak shape.
  • Minimal dependence on strong ion pairing agents, ideal for LC or LC-MS.
  • Ultra-low dispersion hardware for exceptional separation, robustness, and high throughput in HPLC.
  • Seamless transition path toward UPLC adoption.
  • Includes column and Cytochrome C Digestion Standard for immediate benchmarking.
  • Digestion standard batches under strict quality control with certificate of analysis.