Learn More
Waters Corp Column, Peptide CSH C18, 2.5 μm, 4.6 × 50 mm
Catalog No. 502392341 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters CSH130 C18 based particles provide excellent peptide peak shape in FA- or TFA-containing mobile phases and exhibit low dependence on strong ion pairing agents, ideal for LC or LC-MS applications. These 2.5 μm eXtended Performance [XP] columns are packed in ultra-low dispersion hardware, ensuring exceptional separation, robustness, and throughput for HPLC assays and a seamless transition to UPLC.
  • C18 chemistry for reversed-phase separation
  • Hybrid, spherical particles; 2.5 μm size, 130 Å pore diameter
  • pH range of 1 to 11; max pressure 10000 psi (690 Bar)
  • Low silanol activity, endcapped; QC tested for peptides
  • Dimensions: 4.6 mm ID × 50 mm length; for UHPLC systems
Catalog No. 50-239-2341 Supplier Waters Corp Supplier No. 186006946
Add to Cart
Add to Cart
Waters CSH130 C18 based particles provide excellent peptide peak shape in FA- or TFA-containing mobile phases and exhibit low dependence on strong ion pairing agents, ideal for LC or LC-MS applications. These 2.5 μm eXtended Performance [XP] columns are packed in ultra-low dispersion hardware, ensuring exceptional separation, robustness, and throughput for HPLC assays and a seamless transition to UPLC.
  • C18 chemistry for reversed-phase separation
  • Hybrid, spherical particles; 2.5 μm size, 130 Å pore diameter
  • pH range of 1 to 11; max pressure 10000 psi (690 Bar)
  • Low silanol activity, endcapped; QC tested for peptides
  • Dimensions: 4.6 mm ID × 50 mm length; for UHPLC systems