Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XSelect Column Kit, C18, 2.5 μm, 130 Å, 2.1 mm

Catalog No. 502392532 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Column Kit features CSH130 C18 particles with defined positive charges, ensuring excellent peak shape for peptides with FA- or TFA-containing mobile phases. Its performance is largely independent of strong ion pairing agents, making it ideal for LC or LC-MS applications. The kit includes one column and a Cytochrome C Digestion Standard vial for immediate benchmarking and continuous monitoring, with the standard prepared under strict quality control and a certificate of analysis.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Hybrid
  • pH range: 1 pH to 11 pH
  • Low silanol activity, endcapped design
  • Molecular weight range: 1000 to 15000
  • Spherical particle shape, 2.5 μm particle size, 130 Å pore size
  • Inner diameter: 2.1 mm; Length: 5 mm
  • System compatibility: UHPLC
  • Application: Peptide analysis
  • Units per package: 3

Catalog No. 50-239-2532 Supplier Waters Corp Supplier No. 176003072
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This Column Kit features CSH130 C18 particles with defined positive charges, ensuring excellent peak shape for peptides with FA- or TFA-containing mobile phases. Its performance is largely independent of strong ion pairing agents, making it ideal for LC or LC-MS applications. The kit includes one column and a Cytochrome C Digestion Standard vial for immediate benchmarking and continuous monitoring, with the standard prepared under strict quality control and a certificate of analysis.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Hybrid
  • pH range: 1 pH to 11 pH
  • Low silanol activity, endcapped design
  • Molecular weight range: 1000 to 15000
  • Spherical particle shape, 2.5 μm particle size, 130 Å pore size
  • Inner diameter: 2.1 mm; Length: 5 mm
  • System compatibility: UHPLC
  • Application: Peptide analysis
  • Units per package: 3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.