Learn More
Waters Corp XSelect Peptide CSH C18 Column, 130Å, 3.5 μm, 4.6 mm X 100 mm
Supplier: Waters Corp 186006956
This column features 3.5 μm particles with a low and carefully-defined concentration of positive charges, which yield excellent peak shape for peptides using either FA- or TFA-containing mobile phases. Its performance shows minimal dependence on strong ion pairing agents, making it ideal for LC or LC-MS applications.
- CSH particle technology
- C18 chemistry
- Reversed phase separation mode
- Hybrid particle substrate
- pH range of 1 to 11
- Maximum pressure up to 6000 psi (415 bar)
- Endcapped particles
- Low silanol activity
- Molecular weight range of 1000 to 15000
- Spherical particle shape
- 3.5 μm particle size
- 130 Å pore size
- Peptide QC tested
- HPLC system compatible
- L1 USP classification
- 4.6 mm inner diameter
- 100 mm length
- 15% carbon load
- Designed for peptide applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.