Learn More
Waters Corp XSelect Peptide CSH C18 Column, 130Å, 3.5 μm, 2.1 mm × 100 mm
Catalog No. 502392476 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column utilizes charged surface hybrid (CSH) C18 3.5 μm particles with low, carefully-defined positive charges. This design provides excellent peptide peak shapes with FA or TFA mobile phases, and minimal dependence on strong ion pairing agents, making it suitable for LC or LC-MS applications.
  • Delivers superior peptide separations with excellent peak capacities in HPLC, UPLC, or LC-MS.
  • Effective with TFA-containing eluents and weaker acid modifiers like formic acid.
  • Offers enhanced mass loading for complex peptide samples.
  • Features scalable chemistry, supporting nanoflow analysis to lab-scale purification.
  • Ensures reproducibility and quality control through batch testing.
  • Provides improved peptide peak shape with minimal dependence on strong ion pairing agents.
  • Allows increased sample loadability for challenging separations.
Catalog No. 50-239-2476 Supplier Waters Corp Supplier No. 186006951
Add to Cart
Add to Cart
This column utilizes charged surface hybrid (CSH) C18 3.5 μm particles with low, carefully-defined positive charges. This design provides excellent peptide peak shapes with FA or TFA mobile phases, and minimal dependence on strong ion pairing agents, making it suitable for LC or LC-MS applications.
  • Delivers superior peptide separations with excellent peak capacities in HPLC, UPLC, or LC-MS.
  • Effective with TFA-containing eluents and weaker acid modifiers like formic acid.
  • Offers enhanced mass loading for complex peptide samples.
  • Features scalable chemistry, supporting nanoflow analysis to lab-scale purification.
  • Ensures reproducibility and quality control through batch testing.
  • Provides improved peptide peak shape with minimal dependence on strong ion pairing agents.
  • Allows increased sample loadability for challenging separations.