Learn More
Waters Corp HPLC Column Kit, XSelect Peptide CSH C18, 4.6 x 20 mm, 3.5 μm, 130 Å
Supplier: Waters Corp 176003085
These XSelect HPLC columns are designed for chromatographic method development, offering diverse sorbents for challenging analyte separations. Utilizing Ethylene-Bridged Hybrid (BEH) and Charged Surface Hybrid (CSH) particle technologies, they improve sample loadability and peak asymmetry in low-ionic-strength mobile phases, while maintaining mechanical and chemical stability.
- Carbon load: 15.0%.
- C18 (RP18, ODS, Octadecyl) package specification.
- Endcapping is present.
- Inner diameter: 4.6 mm.
- Mode: Reversed Phase (RP).
- Particle size: 3.5 μm.
- pH range: 1 to 11.
- Pore size: 130 Å.
- USP number: L1.
- Length: 20 mm.
- Maximum operating pressure: Up to 6000 psi (400 bar / 40 Mpa).
- Recommended operating temperature: Up to 80°C.
- Each shipped column is individually tested for packed bed efficiency.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.