Learn More
Waters Corp Waters Column, XSelect CSH130 C18, 2.1 x 150 mm, 3.5 μm
Catalog No. 502391046 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect CSH130 C18 Column features Charged Surface Hybrid particles for superior peptide separations in LC or LC–MS. It works with standard TFA–containing or weaker acid eluents, minimizing MS signal reduction. This column accepts greater peptide mass loads, enhancing detection of low–level constituents.
  • Delivers outstanding peak capacities in HPLC, UPLC™, or LC–MS separations of peptides and small proteins up to approximately 10,000 Daltons.
  • Provides excellent mass loading for complex peptide samples, suitable for peptide mapping and proteomics applications.
  • Each batch is QC tested to ensure column–to–column reproducibility and consistent performance.
  • Contains a low concentration of positive charges, yielding excellent peak shape for peptides with FA– or TFA–containing mobile phases.
Catalog No. 50-239-1046 Supplier Waters Corp Supplier No. 176003079
The XSelect CSH130 C18 Column features Charged Surface Hybrid particles for superior peptide separations in LC or LC–MS. It works with standard TFA–containing or weaker acid eluents, minimizing MS signal reduction. This column accepts greater peptide mass loads, enhancing detection of low–level constituents.
  • Delivers outstanding peak capacities in HPLC, UPLC™, or LC–MS separations of peptides and small proteins up to approximately 10,000 Daltons.
  • Provides excellent mass loading for complex peptide samples, suitable for peptide mapping and proteomics applications.
  • Each batch is QC tested to ensure column–to–column reproducibility and consistent performance.
  • Contains a low concentration of positive charges, yielding excellent peak shape for peptides with FA– or TFA–containing mobile phases.