Learn More
Waters Corp XSelect Peptide CSH C18 Column Kit, 130Å, 3.5 μm, 4.6 mm × 50 mm
Catalog No. 502391058 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The particles in this column feature a low and precisely defined concentration of positive charges. This design results in excellent peak shape for peptides, whether using FA- or TFA-containing mobile phases. The column’s performance is minimally affected by strong ion pairing agents, making it well-suited for both LC and LC-MS applications.
  • Designed with a low and precisely defined concentration of positive charges for excellent peak shape for peptides.
  • Provides consistent performance with both FA- or TFA-containing mobile phases.
  • Exhibits minimal dependence on strong ion pairing agents, ideal for LC or LC-MS applications.
  • Kit includes a column and a vial of Cytochrome C Digestion Standard for immediate benchmarking.
  • Digestion standard batches are prepared under strict quality control, confirmed by an included certificate of analysis.
Catalog No. 50-239-1058 Supplier Waters Corp Supplier No. 176003082
The particles in this column feature a low and precisely defined concentration of positive charges. This design results in excellent peak shape for peptides, whether using FA- or TFA-containing mobile phases. The column’s performance is minimally affected by strong ion pairing agents, making it well-suited for both LC and LC-MS applications.
  • Designed with a low and precisely defined concentration of positive charges for excellent peak shape for peptides.
  • Provides consistent performance with both FA- or TFA-containing mobile phases.
  • Exhibits minimal dependence on strong ion pairing agents, ideal for LC or LC-MS applications.
  • Kit includes a column and a vial of Cytochrome C Digestion Standard for immediate benchmarking.
  • Digestion standard batches are prepared under strict quality control, confirmed by an included certificate of analysis.